Gene Transcripts

Gene Symbol
Nanos1
Entrez Gene ID
332397
Gene Name
nanos homolog 1 (Drosophila)
Gene Aliases
Chromosome Location
Chr. 19: 60755987 - 60759914 on Build GRCm38
UniGene ID
Mm.186390
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178421.3 NP_848508.2 1 1916
GenBank mRNA AB095029.1 - 1 1496
AK046189.1 - 1 1520
AK139148.1 - 1 1726
BC056473.1 - 1 1603
BC085512.1 - 1 1916 6849859

View Full Product Details