Gene Transcripts

Gene Symbol
Slitrk3
Entrez Gene ID
386750
Gene Name
SLIT and NTRK-like family, member 3
Gene Aliases
Chromosome Location
Chr. 3: 73047247 - 73057881 on Build GRCm38
UniGene ID
Mm.331076
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198864.2 NP_942564.2 2 556
GenBank mRNA AK139378.1 - 2 680
AK139438.1 - 2 333
AK158856.1 - 2 411
BC066059.1 - 2 556 6844461

View Full Product Details