Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145383.1 | NP_663358.1 | 1 | 364 | |
GenBank mRNA | AK044333.1 | - | 1 | 384 | |
AK044412.1 | - | 1 | 382 | ||
BC013125.1 | - | 1 | 364 | 4500760 | |
BC031766.1 | - | 1 | 365 | 4527040 |