Gene Transcripts

Gene Symbol
Mfsd7c
Entrez Gene ID
217721
Gene Name
major facilitator superfamily domain containing 7C
Gene Aliases
CCT, Flvcr2
Chromosome Location
Chr. 12: 85746539 - 85813585 on Build GRCm38
UniGene ID
Mm.301076
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145447.2 NP_663422.1 6 1523
GenBank mRNA AK145696.1 - 6 1480
AY260574.1 - 6 1523
AY260575.1 - 6 1523
AY260576.1 - 6 1523
BC011209.1 - 6 1523 4190027

View Full Product Details