Gene Transcripts

Gene Symbol
Ubxn1
Entrez Gene ID
225896
Gene Name
UBX domain protein 1
Gene Aliases
2B28, 4930455J02Rik, D19Ertd721e, T25529
Chromosome Location
Chr. 19: 8871510 - 8875664 on Build GRCm38
UniGene ID
Mm.27839
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146093.1 NP_666205.1 8, 9 698
GenBank mRNA AK150442.1 - 7, 8 701
AK159708.1 - 7, 8 673
BC006701.1 - 8, 9 698 3583916

View Full Product Details