Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145349.1 | NP_001138821.1 | 2 | 410 | |
NM_005897.2 | NP_005888.1 | 2 | 410 | ||
XM_005270839.4 | XP_005270896.1 | 2 | 410 | ||
XM_006710623.3 | XP_006710686.1 | 2 | 410 | ||
XM_011541394.2 | XP_011539696.1 | 2 | 410 | ||
XM_017001210.1 | XP_016856699.1 | 2 | 410 | ||
GenBank mRNA | AF156857.1 | - | 1 | 193 | |
AK023880.1 | - | 2 | 246 | ||
BC032544.1 | - | 2 | 251 | 5535760 | |
DA905423.1 | - | 2 | 412 |