Gene Transcripts

Gene Symbol
C330027C09Rik
Entrez Gene ID
224171
Gene Name
RIKEN cDNA C330027C09 gene
Gene Aliases
AA408511, AU018569, Cip2a, Kiaa1524
Chromosome Location
Chr. 16: 48994188 - 49019711 on Build GRCm38
UniGene ID
Mm.24491
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172616.2 NP_766204.2 1 136
GenBank mRNA AK049356.1 - 1 138
AK160639.1 - 1 122
BC137719.1 - 1 145 8860738

View Full Product Details