Gene Transcripts

Gene Symbol
Ctso
Entrez Gene ID
229445
Gene Name
cathepsin O
Gene Aliases
A330105D01Rik, A430056C07, AI118514
Chromosome Location
Chr. 3: 81932616 - 81956725 on Build GRCm38
UniGene ID
Mm.254642
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177662.2 NP_808330.1 2 186
GenBank mRNA AK034490.1 - 2 186
AK040071.1 - 2 160
AK049470.1 - 2 159
AK143683.1 - 2 202
AK165930.1 - 2 148
AK166103.1 - 2 155
BC044664.1 - 2 78
BC098219.1 - 2 160 1066724

View Full Product Details