Gene Transcripts

Gene Symbol
Gprc5a
Entrez Gene ID
232431
Gene Name
G protein-coupled receptor, family C, group 5, member A
Gene Aliases
Rai3, Raig1
Chromosome Location
Chr. 6: 135065662 - 135084709 on Build GRCm38
UniGene ID
Mm.23575
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC034063.1 - 3 720
BC036173.1 - 4 1217 4989149
BC036174.1 - 3 997
BC039217.1 - 4 1169 4205657

View Full Product Details