Gene Transcripts

Gene Symbol
Ccdc85a
Entrez Gene ID
216613
Gene Name
coiled-coil domain containing 85A
Gene Aliases
Chromosome Location
Chr. 11: 28385684 - 28584441 on Build GRCm38
UniGene ID
Mm.93759
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181577.5 NP_853555.2 3, 4 2083
GenBank mRNA AK045568.1 - 3, 4 2056
BC020949.1 - 2, 3 1046

View Full Product Details