Gene Transcripts

Gene Symbol
Adam33
Entrez Gene ID
110751
Gene Name
a disintegrin and metallopeptidase domain 33
Gene Aliases
Adaml
Chromosome Location
Chr. 2: 131050817 - 131063814 on Build GRCm38
UniGene ID
Mm.108550
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163529.1 NP_001157001.1 4, 5 638
NM_033615.2 NP_291093.2 4, 5 638
GenBank mRNA AB059632.1 - 4, 5 638
AB059633.1 - 4, 5 638
AF472524.1 - 4, 5 370
AY054984.1 - 4, 5 338
AY382193.1 - 4, 5 369
AY382194.1 - 4, 5 369
BC138139.1 - 4, 5 361 8861158

View Full Product Details