Gene Transcripts

Gene Symbol
Rnf144a
Entrez Gene ID
108089
Gene Name
ring finger protein 144A
Gene Aliases
Rnf144, UIP4, Ubce7ip4
Chromosome Location
Chr. 12: 26306794 - 26415321 on Build GRCm38
UniGene ID
Mm.214932
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081977.2 NP_001075446.1 10 1940
NM_080563.4 NP_542130.1 9 1866
GenBank mRNA AK038586.1 - 9 1758
AK133544.1 - 7 1480
AK172904.1 - 8 1701
BC030187.1 - 9 1830 5345434

View Full Product Details