Gene Transcripts

Gene Symbol
Slc2a10
Entrez Gene ID
170441
Gene Name
solute carrier family 2 (facilitated glucose transporter), member 10
Gene Aliases
AA450473, Glut10
Chromosome Location
Chr. 2: 165503897 - 165519917 on Build GRCm38
UniGene ID
Mm.34147
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130451.3 NP_569718.1 5 2347
GenBank mRNA AK052156.1 - 5 2314
AY029579.1 - 5 2269

View Full Product Details