Gene Transcripts

Gene Symbol
Rtn4ip1
Entrez Gene ID
170728
Gene Name
reticulon 4 interacting protein 1
Gene Aliases
D10Ertd690e, NIMP
Chromosome Location
Chr. 10: 43901807 - 43947862 on Build GRCm38
UniGene ID
Mm.390253
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130892.4 NP_570962.2 9 2842
GenBank mRNA AK050324.1 - 9 2840
AK088029.1 - 9 2844
BC024116.1 - 9 2812 5100503

View Full Product Details