Gene Transcripts

Gene Symbol
Lacc1
Entrez Gene ID
210808
Gene Name
laccase (multicopper oxidoreductase) domain containing 1
Gene Aliases
9030625A04Rik
Chromosome Location
Chr. 14: 77024201 - 77036898 on Build GRCm38
UniGene ID
Mm.89278
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172488.2 NP_766076.1 3 661
GenBank mRNA AK033565.1 - 3 662
AK053693.1 - 3 946
AK152154.1 - 3 602
AK153349.1 - 3 686
BC116748.1 - 2 417 40126067
BC119140.1 - 2 417 8733889

View Full Product Details