Gene Transcripts

Gene Symbol
Esrp1
Entrez Gene ID
207920
Gene Name
epithelial splicing regulatory protein 1
Gene Aliases
2210008M09Rik, A630065D16, BC031468, Rbm35a
Chromosome Location
Chr. 4: 11331933 - 11386783 on Build GRCm38
UniGene ID
Mm.387719
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290383.1 NP_001277312.1 13 2060
NM_194055.3 NP_918944.2 13 2063
GenBank mRNA AK140848.1 - 13 2061
BC031468.1 - 11 1354 4037840
BC059280.1 - 11 1364

View Full Product Details