Gene Transcripts
- Gene Symbol
- Elp4
- Entrez Gene ID
- 77766
- Gene Name
- elongation protein 4 homolog (S. cerevisiae)
- Gene Aliases
- A330107A17Rik, Paxneb
- Chromosome Location
- Chr. 2: 105700109 - 105904500 on Build GRCm38
- UniGene ID
- Mm.234998
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_023876.3 |
NP_076365.2
|
5, 6 |
657 |
|
GenBank mRNA |
AJ276004.1 |
- |
5, 6 |
656 |
|
AK020742.1 |
- |
5, 6 |
665 |
|
AK034040.1 |
- |
5, 6 |
662 |
|
AK034529.1 |
- |
5, 6 |
663 |
|
AK037104.1 |
- |
5, 6 |
681 |
|
AK039447.1 |
- |
5, 6 |
669 |
|
AK039758.1 |
- |
5, 6 |
658 |
|
AK079069.1 |
- |
5, 6 |
672 |
|
AK168964.1 |
- |
5, 6 |
721 |
|
View Full Product Details