Gene Transcripts

Gene Symbol
Pkd1l2
Entrez Gene ID
76645
Gene Name
polycystic kidney disease 1 like 2
Gene Aliases
1700126L06Rik
Chromosome Location
Chr. 8: 116995218 - 117082488 on Build GRCm38
UniGene ID
Mm.159753
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029686.4 NP_083962.4 1 245
GenBank mRNA AY164484.1 - 1 245

View Full Product Details