Gene Transcripts
- Gene Symbol
- Hdhd2
- Entrez Gene ID
- 76987
- Gene Name
- haloacid dehalogenase-like hydrolase domain containing 2
- Gene Aliases
- 0610039H12Rik, 3110052N05Rik
- Chromosome Location
- Chr. 18: 76944115 - 76972171 on Build GRCm38
- UniGene ID
- Mm.236127
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001039201.1 |
NP_001034290.1
|
2, 3 |
219 |
|
NM_001039202.1 |
NP_001034291.1
|
2, 3 |
180 |
|
NM_029826.2 |
NP_084102.1
|
2, 3 |
180 |
|
GenBank mRNA |
AK002837.1 |
- |
2, 3 |
168 |
|
AK014208.1 |
- |
2, 3 |
174 |
|
AK137285.1 |
- |
2, 3 |
172 |
|
AK147794.1 |
- |
|
201 |
|
AK151285.1 |
- |
2, 3 |
168 |
|
AK152162.1 |
- |
2, 3 |
168 |
|
AK152259.1 |
- |
2, 3 |
168 |
|
BC058177.1 |
- |
2, 3 |
154 |
5250739 |
BY069367.1 |
- |
2, 3 |
190 |
|
BY141971.1 |
- |
3, 4 |
268 |
|
View Full Product Details