Gene Transcripts

Gene Symbol
Cstad
Entrez Gene ID
78617
Gene Name
CSA-conditional, T cell activation-dependent protein
Gene Aliases
9530039B02Rik
Chromosome Location
Chr. 2: 30595044 - 30608945 on Build GRCm38
UniGene ID
Mm.40413
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030137.2 NP_084413.1 2 701
GenBank mRNA AK020589.1 - 2 703
AK047294.1 - 2 702

View Full Product Details