Gene Transcripts

Gene Symbol
Tas1r3
Entrez Gene ID
83771
Gene Name
taste receptor, type 1, member 3
Gene Aliases
Sac, T1r3
Chromosome Location
Chr. 4: 155859268 - 155863353 on Build GRCm38
UniGene ID
Mm.358792
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031872.2 NP_114078.1 6 2249
GenBank mRNA AB049994.1 - 6 2262
AF311386.1 - 6 2230
AF337039.1 - 6 2249
AY032621.1 - 6 2230

View Full Product Details