Gene Transcripts

Gene Symbol
Mycbpap
Entrez Gene ID
104601
Gene Name
MYCBP associated protein
Gene Aliases
4932408B01Rik, AMAP-1, AW125474
Chromosome Location
Chr. 11: 94501347 - 94532388 on Build GRCm38
UniGene ID
Mm.84725
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_170671.2 NP_733771.2 9, 10 1171
GenBank mRNA AK029929.1 - 8, 9 1347
AK166780.1 - 2, 3 319
BC027327.1 - 5, 6 532 3991131

View Full Product Details