Gene Transcripts

Gene Symbol
Wdr92
Entrez Gene ID
103784
Gene Name
WD repeat domain 92
Gene Aliases
AI553587, HZGJ
Chromosome Location
Chr. 11: 17211893 - 17235200 on Build GRCm38
UniGene ID
Mm.298132
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178909.4 NP_849240.1 8 1864
GenBank mRNA AK033294.1 - 8 1810
AK088218.1 - 8 1839
AK142228.1 - 8 1811
BC096540.1 - 8 1794 6403681

View Full Product Details