Gene Transcripts

Gene Symbol
Igfbpl1
Entrez Gene ID
75426
Gene Name
insulin-like growth factor binding protein-like 1
Gene Aliases
2810011G06Rik, 2810453O06Rik, IGFBPL
Chromosome Location
Chr. 4: 45809507 - 45826827 on Build GRCm38
UniGene ID
Mm.3919
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018741.2 NP_061211.1 5 1411
GenBank mRNA AB006141.1 - 5 1399

View Full Product Details