Gene Transcripts

Gene Symbol
Hsdl2
Entrez Gene ID
72479
Gene Name
hydroxysteroid dehydrogenase like 2
Gene Aliases
2610207I16Rik
Chromosome Location
Chr. 4: 59580458 - 59618694 on Build GRCm38
UniGene ID
Mm.272905
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301367.1 NP_001288296.1 12 2057
NM_024255.3 NP_077217.2 11 1959
GenBank mRNA AA545425.1 - 11
AJ293845.1 - 12 1999
AK011910.1 - 11 1907
AK085899.1 - 11 1929
AK136773.1 - 11 1928
AK144948.1 - 11 1912
AK145230.1 - 11 1855
AK208799.1 - 1 149
AY725196.1 - 11 1816
BC002211.1 - 11 1850 3490303

View Full Product Details