Gene Transcripts

Gene Symbol
Cic
Entrez Gene ID
71722
Gene Name
capicua homolog (Drosophila)
Gene Aliases
1200010B10Rik, mKIAA0306
Chromosome Location
Chr. 7: 25267657 - 25294159 on Build GRCm38
UniGene ID
Mm.28833
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB093224.1 - 1 104
AF363690.1 - 6 834
AK147276.1 - 6 1467
AK147535.1 - 7 3585
BC058665.1 - 3 352

View Full Product Details