Gene Transcripts

Gene Symbol
Ift57
Entrez Gene ID
73916
Gene Name
intraflagellar transport 57
Gene Aliases
4833420A15Rik, Esrrbl1, Hippi, MHS4R2
Chromosome Location
Chr. 16: 49699294 - 49765126 on Build GRCm38
UniGene ID
Mm.241276
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028680.3 NP_082956.2 11 1739
GenBank mRNA AK047217.1 - 11 1739
BC138143.1 - 11 1713 8861162
BC138144.1 - 11 1713 8861163

View Full Product Details