Gene Transcripts

Gene Symbol
Agk
Entrez Gene ID
69923
Gene Name
acylglycerol kinase
Gene Aliases
2610037M15Rik, 6720408I04Rik, AI465370, Mulk
Chromosome Location
Chr. 6: 40325155 - 40397562 on Build GRCm38
UniGene ID
Mm.32840
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023538.2 NP_076027.1 14 1048
GenBank mRNA AJ401619.1 - 14 1021
AK076224.1 - 13 1011
AY772469.1 - 13 989
BC019145.1 - 14 1042 5051535
BC093525.1 - 14 1018 5151866

View Full Product Details