Gene Transcripts

Gene Symbol
Agk
Entrez Gene ID
69923
Gene Name
acylglycerol kinase
Gene Aliases
2610037M15Rik, 6720408I04Rik, AI465370, Mulk
Chromosome Location
Chr. 6: 40325155 - 40397562 on Build GRCm38
UniGene ID
Mm.32840
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023538.2 NP_076027.1 16 1966
GenBank mRNA AJ401619.1 - 16 1940
AK076224.1 - 16 1869
BC019145.1 - 16 1960 5051535
BC093525.1 - 16 1936 5151866

View Full Product Details