Gene Transcripts

Gene Symbol
Mul1
Entrez Gene ID
68350
Gene Name
mitochondrial ubiquitin ligase activator of NFKB 1
Gene Aliases
0610009K11Rik, AV000801, Gide
Chromosome Location
Chr. 4: 138434661 - 138442265 on Build GRCm38
UniGene ID
Mm.103413
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026689.3 NP_080965.2 4 2419
GenBank mRNA AK076419.1 - 4 2380
AK083295.1 - 4 2391
AK153956.1 - 4 2369

View Full Product Details