Gene Transcripts

Gene Symbol
Kbtbd3
Entrez Gene ID
69149
Gene Name
kelch repeat and BTB (POZ) domain containing 3
Gene Aliases
2200003A07Rik, Bklhd3
Chromosome Location
Chr. 9: 4309743 - 4331829 on Build GRCm38
UniGene ID
Mm.25946
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164574.1 NP_001158046.1 3 655
NM_026962.3 NP_081238.2 4 687
GenBank mRNA AK034334.1 - 4 595
AK078744.1 - 3 567
AK083497.1 - 4 669
BC027392.1 - 4 591 4954712

View Full Product Details