Gene Transcripts
- Gene Symbol
- Tmem30a
- Entrez Gene ID
- 69981
- Gene Name
- transmembrane protein 30A
- Gene Aliases
- 2010200I23Rik, AW540225, Cdc50a, D9Wsu20e
- Chromosome Location
- Chr. 9: 79760276 - 79794028 on Build GRCm38
- UniGene ID
- Mm.486324
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_133718.4 |
NP_598479.1
|
4, 5 |
758 |
|
GenBank mRNA |
AK008408.1 |
- |
4, 5 |
644 |
|
AK077065.1 |
- |
4, 5 |
761 |
|
AK150071.1 |
- |
4, 5 |
661 |
|
AK159364.1 |
- |
4, 5 |
685 |
|
AK161096.1 |
- |
4, 5 |
640 |
|
AK170171.1 |
- |
4, 5 |
661 |
|
AK170697.1 |
- |
4, 5 |
662 |
|
BC018367.1 |
- |
4, 5 |
610 |
2648510 |
BC018491.1 |
- |
4, 5 |
638 |
2651602 |
BC026136.1 |
- |
3, 4 |
234 |
|
View Full Product Details