Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164566.1 | NP_001158038.1 | 3 | 217 | |
NM_144882.4 | NP_659131.2 | 5 | 403 | ||
GenBank mRNA | AK012791.1 | - | 5 | 377 | |
AK019292.1 | - | 5 | 377 | ||
AK031266.1 | - | 5 | 358 | ||
AK032169.1 | - | 5 | 338 | ||
BC014764.1 | - | 3 | 217 | 4217648 | |
BC021879.1 | - | 5 | 339 | 4216816 | |
BY069430.1 | - | 3 | 173 | ||
BY739337.1 | - | 3 | 180 | ||
EF410080.1 | - | 443 |