Gene Transcripts

Gene Symbol
Slc44a2
Entrez Gene ID
68682
Gene Name
solute carrier family 44, member 2
Gene Aliases
1110028E10Rik, CTL2
Chromosome Location
Chr. 9: 21320719 - 21355028 on Build GRCm38
UniGene ID
Mm.148425
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199186.1 NP_001186115.1 20 2004
NM_152808.3 NP_690021.1 20 2006
GenBank mRNA AK031021.1 - 1 1622
AK036249.1 - 6 2342
AK041533.1 - 20 1963
AK048648.1 - 20 1991
AK153992.1 - 20 1977
AK166695.1 - 20 2007
BC013213.1 - 13 1399
BC023763.1 - 14 1241
BC031535.1 - 20 1982 3991609

View Full Product Details