Gene Transcripts
- Gene Symbol
- Psmd11
- Entrez Gene ID
- 69077
- Gene Name
- proteasome (prosome, macropain) 26S subunit, non-ATPase, 11
- Gene Aliases
- 1700089D09Rik, 1810019E17Rik, 2610024G20Rik, 2810055C24Rik, C78232, P44.5, S9
- Chromosome Location
- Chr. 11: 80428615 - 80473248 on Build GRCm38
- UniGene ID
- Mm.260539
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_178616.3 |
NP_848731.2
|
3, 4 |
373 |
|
GenBank mRNA |
AK007547.1 |
- |
3, 4 |
342 |
|
AK012951.1 |
- |
3, 4 |
382 |
|
AK051738.1 |
- |
3, 4 |
363 |
|
AK076002.1 |
- |
3, 4 |
361 |
|
AK089942.1 |
- |
3, 4 |
338 |
|
AK136049.1 |
- |
3, 4 |
336 |
|
AK140773.1 |
- |
|
357 |
|
AK167559.1 |
- |
3, 4 |
344 |
|
AK168289.1 |
- |
3, 4 |
365 |
|
AK192313.1 |
- |
1, 2 |
56 |
|
BC030432.1 |
- |
|
27 |
|
BC037590.1 |
- |
3, 4 |
322 |
|
BC055457.1 |
- |
3, 4 |
305 |
|
BC090664.1 |
- |
3, 4 |
325 |
30677672 |
BC090980.1 |
- |
3, 4 |
340 |
30283128 |
BC100595.1 |
- |
3, 4 |
335 |
6767671 |
BC119136.1 |
- |
3, 4 |
344 |
8733885 |
BC119138.1 |
- |
3, 4 |
344 |
8733887 |
View Full Product Details