Gene Transcripts

Gene Symbol
Sec61a2
Entrez Gene ID
57743
Gene Name
Sec61, alpha subunit 2 (S. cerevisiae)
Gene Aliases
Chromosome Location
Chr. 2: 5870977 - 5895427 on Build GRCm38
UniGene ID
Mm.21435
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021305.3 NP_067280.1 5 414
GenBank mRNA AF145254.1 - 5 350
AF222748.1 - 5 351
AK017615.1 - 5 396
AK077701.1 - 5 414
AK136447.1 - 5 393
AK160602.1 - 5 400
AK161316.1 - 5 493
BC005458.1 - 5 357 3494001

View Full Product Details