Gene Transcripts

Gene Symbol
Acer3
Entrez Gene ID
66190
Gene Name
alkaline ceramidase 3
Gene Aliases
1110057L18Rik, 5430429L08Rik, AV015045, Phca
Chromosome Location
Chr. 7: 98213660 - 98309546 on Build GRCm38
UniGene ID
Mm.334041
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025408.2 NP_079684.2 1 168
GenBank mRNA AK004287.1 - 1 169
AK011668.1 - 1 149
AK080977.1 - 1 146
AK150629.1 - 1 129
AK162403.1 - 1 184
BC023924.1 - 1 172 5324078

View Full Product Details