Gene Transcripts

Gene Symbol
Ier3ip1
Entrez Gene ID
66191
Gene Name
immediate early response 3 interacting protein 1
Gene Aliases
1110057H19Rik, AI644142, AL022842, AL022863, AV026606
Chromosome Location
Chr. 18: 76930027 - 76941614 on Build GRCm38
UniGene ID
Mm.28593
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025409.3 NP_079685.1 2 269
GenBank mRNA AK004281.1 - 2 254
AK007388.1 - 2 217
AK007862.1 - 2 201
AK009116.1 - 2 202
AK190140.1 - 2 140
AK210106.1 - 2 137
BC021512.1 - 2 259 5351700
CX207811.1 - 2 258

View Full Product Details