Gene Transcripts

Gene Symbol
Eapp
Entrez Gene ID
66266
Gene Name
E2F-associated phosphoprotein
Gene Aliases
1810011O16Rik
Chromosome Location
Chr. 12: 54670340 - 54695896 on Build GRCm38
UniGene ID
Mm.156440
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025456.3 NP_079732.1 4 472
GenBank mRNA AK007442.1 - 4 427
AK012800.1 - 4 429
AK145027.1 - 4 491
AK145041.1 - 4 491
AY882557.1 - 4 391
BC037622.1 - 4 499 5373275

View Full Product Details