Gene Transcripts

Gene Symbol
Lsm14a
Entrez Gene ID
67070
Gene Name
LSM14 homolog A (SCD6, S. cerevisiae)
Gene Aliases
2700023B17Rik, AA407828, AU017544, Tral
Chromosome Location
Chr. 7: 34344637 - 34389637 on Build GRCm38
UniGene ID
Mm.276503
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025948.2 NP_080224.1 2 218
GenBank mRNA BC031521.1 - 2 218

View Full Product Details