Gene Transcripts

Gene Symbol
Cdt1
Entrez Gene ID
67177
Gene Name
chromatin licensing and DNA replication factor 1
Gene Aliases
2610318F11Rik, AW545653, C76791, Ris2
Chromosome Location
Chr. 8: 122568015 - 122573130 on Build GRCm38
UniGene ID
Mm.21873
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026014.3 NP_080290.3 5 870
GenBank mRNA AB086655.1 - 5 849
AF477481.1 - 5 870
AK012041.1 - 5 847
AK017609.1 - 5 844
AK028287.1 - 5 842
AK049163.1 - 5 842
AK076527.1 - 4 952
AK083015.1 - 5 845
AK208010.1 - 1 43
BC013842.1 - 5 825
BC024485.1 - 2 121
BC048076.1 - 5 870 6307476

View Full Product Details