Gene Transcripts

Gene Symbol
Hils1
Entrez Gene ID
54388
Gene Name
histone H1-like protein in spermatids 1
Gene Aliases
H1.9, TISP64
Chromosome Location
Chr. 11: 94967661 - 94968456 on Build GRCm38
UniGene ID
Mm.30482
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018792.1 NP_061262.1 1 338
GenBank mRNA AB022320.1 - 1 338
AB045724.1 - 1 331
AK005718.1 - 1 369
AY286319.1 - 1 338
BC050966.1 - 1 371
BC061119.1 - 1 401 6773493

View Full Product Details