Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019780.1 | NP_062754.1 | 2 | 203 | |
GenBank mRNA | AB578902.1 | - | 3 | 213 | |
AB578903.1 | - | 3 | 182 | ||
AB578904.1 | - | 2 | 218 | ||
AB578905.1 | - | 2 | 207 | ||
AB578906.1 | - | 2 | 201 | ||
AB578907.1 | - | 2 | 200 | ||
AB578908.1 | - | 2 | 172 | ||
AB578909.1 | - | 2 | 217 | ||
AB578910.1 | - | 2 | 186 | ||
AB578911.1 | - | 2 | 206 | ||
AF193794.1 | - | 2 | 203 | ||
AK004103.1 | - | 3 | 220 | ||
AK008248.1 | - | 3 | 223 | ||
AK036426.1 | - | 2 | 191 | ||
AK044468.1 | - | 1 | 2525 | ||
AK150330.1 | - | 2 | 213 | ||
BC005663.1 | - | 2 | 192 | 2645432 |