Gene Transcripts

Gene Symbol
Ick
Entrez Gene ID
56542
Gene Name
intestinal cell kinase
Gene Aliases
2210420N10Rik, AI848300, Mrk
Chromosome Location
Chr. 9: 78109192 - 78172114 on Build GRCm38
UniGene ID
Mm.288719
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163780.1 NP_001157252.1 4 690
NM_019987.2 NP_064371.2 4 618
GenBank mRNA AF225918.1 - 4 458
AK087484.1 - 4 541
AK133537.1 - 4 557
AK154185.1 - 4 691
AK206365.1 - 3 271
BC028863.1 - 3 375 4224269

View Full Product Details