Gene Transcripts

Gene Symbol
Nfu1
Entrez Gene ID
56748
Gene Name
NFU1 iron-sulfur cluster scaffold homolog (S. cerevisiae)
Gene Aliases
0610006G17Rik, CGI-33, Hirip5
Chromosome Location
Chr. 6: 87009291 - 87028461 on Build GRCm38
UniGene ID
Mm.23809
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170591.1 NP_001164062.1 6 577
NM_020045.3 NP_064429.2 6 574
GenBank mRNA AJ132616.1 - 6 505
AK002244.1 - 6 523
AK003730.1 - 6 522
AK145558.1 - 6 578
AY335195.1 - 6 481
BC018355.1 - 6 495 3584656

View Full Product Details