Gene Transcripts

Gene Symbol
Tubgcp4
Entrez Gene ID
51885
Gene Name
tubulin, gamma complex associated protein 4
Gene Aliases
4932441P04Rik, 76p, D2Ertd435e, GCP-4
Chromosome Location
Chr. 2: 121170454 - 121198770 on Build GRCm38
UniGene ID
Mm.383499
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290824.1 NP_001277753.1 12 1479
NM_153387.3 NP_700436.1 12 1479
GenBank mRNA AK034694.1 - 3 1009
AK038921.1 - 12 1438
AK045305.1 - 12 1372
AK046132.1 - 12 1409
AK050250.1 - 12 1480
AK051517.1 - 12 1380
AK152984.1 - 12 1438
AK153171.1 - 12 1438
AK161774.1 - 3 275
AK181112.1 - 4 317
BC029106.1 - 12 1443 3711922

View Full Product Details