Gene Transcripts

Gene Symbol
Zfp42
Entrez Gene ID
22702
Gene Name
zinc finger protein 42
Gene Aliases
Rex-1, Rex1, Zfp-42
Chromosome Location
Chr. 8: 43295067 - 43307009 on Build GRCm38
UniGene ID
Mm.285848
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009556.3 NP_033582.2 4 626
GenBank mRNA AB221555.1 - 1 225
BC053399.1 - 4 599 30031349
BC064785.1 - 5 658
BC098467.1 - 4 561 30008039
M28382.1 - 4 587

View Full Product Details