Gene Transcripts

Gene Symbol
Zfp93
Entrez Gene ID
22755
Gene Name
zinc finger protein 93
Gene Aliases
Znf235
Chromosome Location
Chr. 7: 24270418 - 24277794 on Build GRCm38
UniGene ID
Mm.459431
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009567.4 NP_033593.1 4 975
GenBank mRNA AK028905.1 - 4 813
AK040325.1 - 4 906
AK132229.1 - 4 976
AK146095.1 - 4 966
AK165630.1 - 4 936
BC003776.1 - 698 3496816
BC061487.1 - 3 756 5349153
U46186.1 - 4 967

View Full Product Details