Gene Transcripts

Gene Symbol
Tnfrsf17
Entrez Gene ID
21935
Gene Name
tumor necrosis factor receptor superfamily, member 17
Gene Aliases
BCM, BCMA, Tnfrsf13, Tnfrsf13a
Chromosome Location
Chr. 16: 11313174 - 11320074 on Build GRCm38
UniGene ID
Mm.12935
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011608.1 NP_035738.1 3 789
GenBank mRNA AF061505.1 - 3 789
AK020247.1 - 3 787
BC027519.1 - 3 660 1349977

View Full Product Details