Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011718.2 | NP_035848.1 | 5 | 1180 | |
GenBank mRNA | AK029284.1 | - | 4 | 623 | |
BC114969.1 | - | 6 | 960 | ||
L33719.1 | - | 1 | 115 | ||
U20658.1 | - | 5 | 995 | ||
U30464.1 | - | 5 | 1178 | ||
U61970.1 | - | 4 | 913 | ||
U61971.1 | - | 4 | 625 |